Please login to your online account to display your discounted pricing

Promega™ GloMax™ Multiplus Detection Systems with Instinct™ Software


To measure fluorescence intensity, luminescence or absorbance in a flexible, modular configuration

Manufacturer: promega™  E9032

 View more versions of this product

Catalog No. PRE9032

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Type Base instrument with heating and shaking
For Use With (Application) To measure fluorescence intensity, luminescence or absorbance in a flexible, modular configuration

  • Detection modes include Luminescence and UV-Visible Absorbance
  • Accepts 6-, 12-, 24-, 48-, 96- and 384-well plates
  • Performance through touch screen
  • Data can be saved on instrument and moved via included USB flash drive
  • Works alone as standalone workstation in laboratory, freeing computing resources from data capture
  • Made up of base unit available in two different formats, one with shaking and one with heating and shaking

Luminescence Module:

  • Head-on photon-counting photomultiplier tube (PMT)
  • Detect 3 × 10-21 moles of luciferase, covering a dynamic range over 8 logs
  • Dual-masking system
UV-Visible Absorbance Module:
  • 6-position filter wheel that includes filters for measuring 260, 280, 450, 560, 600 and 750nm
  • Filters accommodate UV DNA and protein quantitation in addition to ELISA and protein assays
  • Customizable by substituting a filter of choice into either of two removable filter paddles

E8032 and E9032 cannot be sold separately and must be purchased with at least one detection module.

Reporter, Cell viability, Apoptosis, Protease, Kinase, ADME, Protein, and ELISA assays. Fluorescence Module: Application-optimized Optical Kits simplify fluorescence operation while maximizing performance. Long-lived LED-based excitation lights minimize maintenance and variability in intensity. LED usage increases sensitivity by fully exciting the fluorophore and reducing nonspecific light leakage, a problem often found when using broad-spectrum light sources. The UV, Blue, Green, Red and AFC Optical Kits are included with the Fluorescence Detection Module.
UV (Ex: 365nm, Em: 410–460nm)
Blue (Ex: 490nm, Em: 510–570nm)
Green (Ex: 525nm, Em: 580–640nm)
Red (Ex: 625nm, Em: 660–720nm)
AFC (Aminofluorocoumarin; Ex: 405nm, Em: 495–505nm)