Please login to your online account to display your discounted pricing

Promega™ GloMax™-Multi Jr Single-Tube Multimode Reader: Optical Kits


Modular system is easy to use, affordable and can be customized to meet a laboratory's application needs

Manufacturer: promega™  E6074

 View more versions of this product

Catalog No. PRE6074

  • / Each
 

Description & Specifications

Specifications

Color Red
For Use With (Equipment) Single-Tube Multimode Reader