Please login to your online account to display your discounted pricing

Promega™ GloMax™-Multi Jr Single-Tube Multimode Reader: Base Instr., Lum. Module, Mod. Serv. Upgr.

Modular system is easy to use, affordable and can be customized to meet a laboratory's application needs

$7,670.83 - $7,670.83

Specifications

For Use With (Application) Luminescence, fluorescence, and absorbance measurements
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

PRE6080

 
promega™
E6080
GloMax-Multi Jr with Luminescence Module Each for $7,670.83
Description & Specifications

Specifications

For Use With (Application) Luminescence, fluorescence, and absorbance measurements

  • Touch screen interface has been designed to be intuitive so that no training is required to use the instrument
  • Easy protocol setup, plus Promega protocols are preloaded for easy implementation
  • Convenient data handling—record data right from the instrument or export data to an Excel™ spreadsheet
  • Small footprint saves bench space

GloMax-Multi Jr with Luminescence Module

  • Designed to deliver performance equivalent to dedicated single-tube luminometers while also offering the flexibility of a multimode reader
  • Sensitivity of 1 × 10-18 moles of luciferase
  • >5 logs of dynamic range, which is more than adequate to cover common luminescence applications, thus reducing the need to dilute samples
Fluorescence Module
  • Designed to deliver both high performance and user flexibility
  • To achieve high performance, each Fluorescence Module utilizes powerful light-emitting diodes (LEDs) as excitation sources
  • LED usage increases sensitivity by fully exciting the fluorophore and reducing nonspecific light leakage, a problem often found when using broad-spectrum light sources
  • Four standard fluorescence optical kits are available: UV (Excitation: 365nm, Emission: 410–450nm), Blue (Excitation: 460nm, Emission: 515–570nm), Green (Excitation: 525nm, Emission: 580–640nm), Red (Excitation: 625nm, Emission: 660–725nm)
  • Custom optical kits are available, contact your Fisher Scientific™ Customer Service Representative for details
Luminescence Module Service Upgrade
  • Permits the addition of the Luminescence Module to the Base Unit (Mfr. No. E6070)
  • Unit must be returned to Promega Service Center for factory installation