missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GloMax™-Multi Jr Single-Tube Multimode Reader: Absorbance Filter Paddle
The GloMax-Multi Jr Single-Tube Multimode Reader is designed to provide the utmost flexibility. In addition to high performance, the GloMax-Multi Jr blends user-friendly operation and a small footprint with flexible purchasing options.
$854.41 - $886.77
Specifications
For Use With (Equipment) | GloMax-Multi Jr Single-Tube Multimode Reader |
---|---|
Includes | 1/ea |
Storage Requirements | 5 to 40°C |
Description
- For Use With the GloMax(R)-Multi Jr Single-Tube Multimode Reader
- Instrument is discontinued
- Accessories (pending availability) and Service Agreement available until November 1, 2020
Specifications
GloMax-Multi Jr Single-Tube Multimode Reader | |
1/ea | |
5 to 40°C |