Promega GloMax™-Multi Detection System: Luminescence Module
An advanced photon-counting photomultiplier tube (PMT) provides unmatched signal-to-noise ratios, beating most standalone luminometers. Promega GloMax Luminescence Module has a dual masking system which minimizes well-to-well cross-talk.
Supplier: Promega E7041
Description
- Detects as little as 3 × 10-21 moles of luciferase, covering a dynamic range over 8 logs
Specifications
Luminescence Module | |
Photomultiplier Tube | |
3 + 10-21 moles of luciferase | |
Reporter assays, Cell viability assays, Apoptosis assays, Protease assays, Kinase assays, ADME assays |
Touchscreen | |
Luminescence | |
350 to 650nm, Eight logs | |
GloMax-Multi Detection System |