missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GloMax™-Multi Detection System: Fluorescence Module
Application-optimized Optical Kits simplify fluorescence operation while maximizing performance. Promega Fluorescence Module for GloMax-Multi Detection System have long-lived LED-based excitation lights which minimize maintenance and variability in intensity.
Supplier: Promega E7051
Includes: UV, Blue, Green, Red and AFC Optical Kits
Description
UV, Blue, Green, Red and AFC Optical Kits are included with the Fluorescence Detection Module.
- UV (Ex: 360nm, Em: 410 to 460nm)
- Blue (Ex: 490nm, Em: 510 to 570nm)
- Green (Ex: 525nm, Em: 580 to 640nm)
- Red (Ex: 625nm, Em: 660 to 720nm)
- AFC (Aminofluorocoumarin; Ex: 405nm, Em: 495 to 505nm)
Specifications
Fluorescence Module | |
Wavelength matched LED | |
Fluorescence | |
UV: 365nm, Blue: 490nm, Green: 525nm, Red: 625nm | |
6 decades (assay dependent) | |
GloMax-Multi Detection System |
Touchscreen | |
PiN-photodiode | |
0.5fmol/200μL, or 1ppt of fluorescein in 96-well microplate | |
UV: 410 to 460nm, Blue: 510 to 570nm, Green: 580 to 640nm, Red: 660 to 720nm | |
Reporter assays, Cell viability assays, Apoptosis assays, Protease assays, Kinase assays, ADME assays | |
UV, Blue, Green, Red and AFC Optical Kits |