missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GloMax™-Multi Detection System: Absorbance Module
A six-position filter wheel with two open positions ensures flexibility for a wide range of applications. Promega Absorbance Module for GloMax-Multi Detection System has an LED-based visible spectrum light source which minimizes maintenance and variability.
Supplier: Promega E7061
Includes: Filters for 450, 560, 600 and 750nm
Description
- Includes filters for reading 450, 560, 600 and 750nm
Specifications
Absorbance Module | |
LED | |
Absorbance | |
Up to six filters: four installed filters and two empty filter holders for user configuration | |
GloMax-Multi Detection System |
Touchscreen | |
Large-area Photodiode | |
0 to 4.0 OD | |
Reporter assays, Cell viability assays, Apoptosis assays, Protease assays, Kinase assays, ADME assays | |
Filters for 450, 560, 600 and 750nm |