Please login to your online account to display your discounted pricing

Promega GloMax™ 96 Microplate Luminometers

Achieve high sensitivity and broad dynamic range. GloMax 96 Microplate Luminometer is a compact, easy to use, low cost luminometer which meets the need of all chemiluminescent and bioluminescent applications.

Specifications

Detection Method Luminescence
View More Specs
Includes:  Power cable, data cable, Quick Protocol card, five white 96-well microplates, and software
Products
Catalog Number Mfr. No. Product Type Price Quantity    

PRE6501

 
promega corportaion
E6501
Microplate Luminometer CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PRE6521

 
promega corportaion
E6521
Microplate Luminometer w/dual injector CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PRE6511

 
promega
E6511
Microplate Luminometer w/single injector CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Detection Method Luminescence

The GloMax 96 is a state-of-the-art microplate luminometer that meets the needs for high sensitivity and broad dynamic range for all luminescence applications.
  • Performs both flash- and glow-type luminescence assays
  • Available with up to two reagent injectors
  • High Sensitivity: Sensitive to approximately 3 x 10-21 moles of luciferase
  • Wide Dynamic Range: 9-log dynamic range
  • Convenient Data Handling: Direct-to-Excel data importing requires Windows™ PC to operate
  • Simple Data Analysis: Excel macros allow simple data analysis for Dual-Luciferase™ assays
  • Advanced photon-counting photomultiplier tube (PMT) provide unmatched signal-to-noise ratios
  • Ideal for cell-based assays
  • Engineered to minimize sample cross-talk
Computer Requirements
The GloMax 96 software is compatible on 32-bit and 64-bit PCs using Windows 7 Professional Service Pack 1 and Microsoft™ Office 2010 Home and Business Edition, 32-bit version. It is also compatible on 64-bit PCs using Windows 8.1 with Microsoft Office 2013. The computer can be connected to the instrument using either an RS-232 cable or USB/RS-232 adaptor.
Applications
Reporter assays, Cell viability assays, Apoptosis assays, Protease assays, Kinase assays, ADME assays

CE

One year