Please login to your online account to display your discounted pricing
Promega™ GloMax™ 20/20 Luminometry System
Touchscreen interface simplifies operation
Manufacturer: promega™  E5311
 View more versions of this product
Catalog No. PRE5311
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Range >8 decades
Holds 1.5mL microcentrifuge tubes and 35mm Petri dishes
Product Type Without injectors