Please login to your online account to display your discounted pricing

Promega™ Genomic DNA


Suitable for Southern blot hybridizations, genomic analysis, and genomic library construction

Manufacturer: promega™  G1521

 View more versions of this product

 Disclaimers

PR-G1471, PR-G1521, PR-G3041 For Laboratory Use

Catalog No. PR-G1521

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Southern blotting hybridizations, genomic analysis (including PCR) and genomic library construction
Product Type Genomic DNA
Quantity 100μg
Source Female Human
Storage Requirements 4°C
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA
Size 100μg