Please login to your online account to display your discounted pricing

Promega™ GeneClip™ U1 Hairpin Cloning Systems


Designed for fast and easy cloning of human hairpin target sequences to allow expression of siRNA target sequences in human cells

Manufacturer: promega™  C8780

 View more versions of this product

Catalog No. PR-C8780

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Fast and easy cloning of Human hairpin target sequences to allow expression of siRNA target sequences in Human cells
Format Liquid
pH 7.4
Format Liquid
Includes 200μL 2X rapid Ligation Buffer, 1.25mL Nuclease-free Water, 1mL Oligo annealing Buffer, 1.2μg pGeneClip Puromycin vector, 100U T4 DNA Ligase
Quantity 20 Reactions
Quantity 20 Reactions