Please login to your online account to display your discounted pricing

Gel Shift Assay Systems


Simple, rapid systems for detecting DNA-binding proteins

Manufacturer: promega corportaion  E3050

 View more versions of this product

Catalog No. PR-E3050

  • / Each
 

Description & Specifications

Specifications

Storage Requirements HeLa nuclear extract at -70°C and other components at -20°C
Includes HeLaScribe™ nuclear extract, 40μL Gel shift assay grade, T4 Polynucleotide Kinase, 100U Kinase Buffer (10X), 100μL Gel shift Binding Buffer (5X), 200μL, AP2 and SP1 Consensus Oligonucleotides, 35pmol each.
Quantity 100 Reactions
Product Type Gel Shift Assay Core System
For Use With (Application) Used to study sequence specific DNA binding Proteins such as transcription facto rs