Please login to your online account to display your discounted pricing

Promega™ Gel Shift Assay Systems DFS Item


Simple, rapid systems for detecting DNA-binding proteins

Manufacturer: promega™  E3300

 View more versions of this product

Catalog No. PR-E3300

  • / Each
 

Description & Specifications

Specifications

Storage Requirements HeLa nuclear extract at -70°C and other components at -20°C
Includes HeLaScribe™ nuclear extract, 40μL Gel shift assay grade, T4 Polynucleotide Kinase, 100U Kinase Buffer (10X), 100μL Gel shift Binding Buffer (5X), 200μL, AP2, SP1, AP1, OCT1, CREB, NF-κB and TFIID Consensus Oligonucleotide, 35pmol each
For Use With (Application) Used to study sequence specific DNA binding Proteins such as transcription facto rs
Product Type Gel Shift Assay Complete System
Quantity 100 Reactions