Please login to your online account to display your discounted pricing
Promega™ Flipper™ Racks
Ideal for storage and transport of all small tubes
Manufacturer: promega™  Y9422
 View more versions of this product
Catalog No. PRY9422
  • / Each
Description & Specifications
Specifications
Holds 0.5mL Microcentrifuge tubes, 1.5mL tubes, 2mL cryogenic tubes
Autoclavable Yes
Positions 96.0
Product Type Rack, Microtube
Autoclavable Yes
Holds 0.5mL Microcentrifuge tubes, 1.5mL tubes, 2mL cryogenic tubes
Material Polypropylene
Material Polypropylene
Type Rack, Microtube
Positions 96.0