Please login to your online account to display your discounted pricing

Promega™ Flexi™ Vectors


For use with Flexi Cloning System

Manufacturer: promega™  C8461

 View more versions of this product

Catalog No. PR-C8461

  • / Each
 

Description & Specifications

Specifications

Quantity 0.02mg
For Use With (Application) Inducible expression of HQ- and GST-tagged fusion Proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
pH 8
Format Fluid
Product Type pFN2A (GST) Flexi Vector
Includes 10X Flexi Enzyme blend (Sgf I and Pme I), 2X flexi Ligase Buffer, 5X flexi Digest Buffer, Nuclease-free Water, T4 DNA Ligase, Wizard™ SV Gel and PCR clean up system (C8640 only)
Concentration 100ng/μL
Concentration 100ng/μL
Storage Requirements -20°C
Quantity 0.02mg
Format Fluid
Product Name pFN2A (GST) Flexi Vector