Please login to your online account to display your discounted pricing

Promega™ Flexi™ Vectors


For use with Flexi Cloning System

Manufacturer: promega™  C8441

 View more versions of this product

Catalog No. PR-C8441

  • / Each
 

Description & Specifications

Specifications

Includes 10X Flexi Enzyme blend (Sgf I and Pme I), 2X flexi Ligase Buffer, 5X flexi Digest Buffer, Nuclease-free Water, T4 DNA Ligase, Wizard™ SV Gel and PCR clean up system (C8640 only)
Storage Requirements -20°C
For Use With (Application) Inducible expression of HQ- and GST-tagged fusion Proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Concentration 100ng/μL
Product Type pF1A T7 Flexi Vector
Quantity 0.02mg
pH 8
Format Fluid