Please login to your online account to display your discounted pricing

Promega™ Flexi™ Vectors

For use with Flexi Cloning System

$352.03 - $398.04

Specifications

Concentration 100ng/μL
Format Fluid
For Use With (Application) Inducible expression of HQ- and GST-tagged fusion Proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Includes 10X Flexi Enzyme blend (Sgf I and Pme I), 2X flexi Ligase Buffer, 5X flexi Digest Buffer, Nuclease-free Water, T4 DNA Ligase, Wizard™ SV Gel and PCR clean up system (C8640 only)
pH 8
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PR-C8441

 
promega™
C8441
Each for $352.03

PR-C8461

 
promega™
C8461
Each for $398.04

PR-L5671

 
promega™
L5671
Each for $398.04
Description & Specifications

Specifications

Concentration 100ng/μL
Format Fluid
For Use With (Application) Inducible expression of HQ- and GST-tagged fusion Proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Includes 10X Flexi Enzyme blend (Sgf I and Pme I), 2X flexi Ligase Buffer, 5X flexi Digest Buffer, Nuclease-free Water, T4 DNA Ligase, Wizard™ SV Gel and PCR clean up system (C8640 only)
pH 8
Storage Requirements -20°C

1L5671, L5681: Licensed under U.S. Pat. No. 5,910,628.
2For research use only. Persons wishing to use this product or its derivatives in other fields of use, including without limitation, commercial sale, diagnostics or therapeutics, should contact Promega Corporation for licensing information.
3This product or portions thereof is manufactured under license from Amrad Corporation Limited. For noncommercial research use only. All other uses require a license from Amrad Corporation Limited, Unit 6, 663 Victoria Street, Abbotsford, Victoria 3067, Australia, under U.S. Patent 5,654,176; Australian Patent 607511, Canadian Patent 1338903 and other issued patents.
4U.S. Patent 5,391,487.
5Licensed under U.S. Patent 5,945,288.
6The “pF” indicates that the vector is a Flexi vector. The letter after “pF” indicates the position of any expression tags (“N” = an N-terminal expression tag). The number associated with the vector specifies the type of expression and application. The letters “A” and “K” designate the bacterial drug selection for the vector (“A” = ampicillin and “K” = kanamycin).