Please login to your online account to display your discounted pricing

Promega™ Flexi™ Enzyme Blends


For use with Flexi Cloning System

Manufacturer: promega™  R1901

 View more versions of this product

 Disclaimers

Licensed under U.S. Patent 5,945,288.

Catalog No. PR1901

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Flexi cloning system
Product Type Enzyme Blend
Format Liquid
pH 7.4
Quantity 0.05mL
Includes Carboxy Flexi™ Enzyme Blend (SgfI & EcoICRI)1 x 50μL, Nuclease-free Water 2 x 1,250μL, 5X Flexi Digest Buffer 1 x 1mL
Quantity 0.05mL
Product Name Enzyme Blend
Format Liquid