Promega FAK Kinase Enzyme System
Recombinant human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. FAK is involved in signal transduction from integrin-enriched focal adhesion sites that mediate cell contact with the extracellular matrix.
Supplier: Promega V1971
Includes:
- 1 x 10μg CSK Kinase Enzyme System (V1971) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Description
- Easily Screen and Profile FAK Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
35 | |
<-65°C | |
10 μg | |
Fluid |