missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Exonuclease III
Exonuclease III is a 3' → 5' exonuclease specific for dsDNA.
Supplier: Promega M1815
Includes: Part No. M1811: Exonuclease III 10X Buffer (1 x 1mL); Exonuclease III (5000U).
Part No. M1815: Exonuclease III 10X Buffer (5 x 1mL); Exonuclease III (25,000U).
Description
- A 3′→5′ Exonuclease Specific for Double-Stranded DNA
- Control deletion rate by varying incubation temperature
- Inactivate Exonuclease III by heating at 75°C for 10 minutes
- Supplied at a concentration of 150-200u/μL
Specifications
150 to 200 U/μL | |
10X Reaction Buffer: 660mM Tris HCl (pH 8 at 25°C), 6.6mM MgCl2 | |
7.5 | |
20mM Tris HCl (pH 7.5 at 25°C), 1mM DTT, 100mM KCl, 50% Glycerol | |
Recombinant E. coli strain | |
Exonuclease III | |
50mM NaCl |
Colorless | |
>90% | |
Generating nested sets of deletions with double-stranded, linear DNA, preparation of single-stranded DNA | |
-30°C to -10°C | |
25,000 U | |
28000 | |
Liquid |