Please login to your online account to display your discounted pricing

Promega™ Emax™ Immunoassay Systems DFS Item

Optimized for the sensitive and specific detection of a variety of neurotrophic and growth factors

$630.23 - $1,225.14

Specifications

Detection Method Chromogenic
Conjugate HRP
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution
Storage Requirements -20°C
View More Specs
Includes:  Antibodies, HRP conjugate, Standard, Buffers, TMB solution
Products
Catalog Number Mfr. No. Quantity Sensitivity Product Type Price Quantity    

PR-G7590

DFS Item
 
promega™
G7590
2 x 96 Well 31pg/mL TGFb1 Emax™ immunoassay system Each for $630.23

PR-G7591

DFS Item
 
promega™
G7591
5 x 96 Well 31pg/mL TGFb2 Emax™ immunoassay system Each for $1,225.14

PR-G7611

DFS Item
 
promega™
G7611
5 x 96 Well 15.6pg/mL BDNF Emax™ immunoassay system Each for $1,225.14

PR-G7620

DFS Item
 
promega™
G7620
2 x 96 Well 31pg/mL GDNF Emax™ immunoassay system Each for $630.23

PR-G7610

DFS Item
 
promega™
G7610
2 x 96 Well 15.6pg/mL BDNF Emax™ immunoassay system Each for $630.23

PR-G7621

DFS Item
 
promega™
G7621
5 x 96 Well 31pg/mL GDNF Emax™ immunoassay system Each for $1,225.14
Description & Specifications

Specifications

Detection Method Chromogenic
Conjugate HRP
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution
Storage Requirements -20°C

  • Detect picogram levels of factor per milliliter of sample using an antibody-sandwich format Utilize a ready-to-use, single-component TMB substrate and horseradish peroxidase (HRP) conjugate formulations that provide increased stability and easier dilutions for final chromogenic detection
  • Provide factor-specific detection with <3% cross-reactivity with other related neurotrophic and growth factors, except TGFβ2 which provides <5% cross-reactivity