Please login to your online account to display your discounted pricing

Promega™ Emax™ Immunoassay Systems DFS Item


Optimized for the sensitive and specific detection of a variety of neurotrophic and growth factors

Manufacturer: promega™  G7620

 View more versions of this product

Catalog No. PR-G7620

  • / Each
 

Description & Specifications

Specifications

Quantity 2 x 96 Well
Conjugate HRP
Product Type GDNF Emax™ immunoassay system
Storage Requirements -20°C
Detection Method Chromogenic
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution
Sensitivity 31pg/mL