Please login to your online account to display your discounted pricing

Promega™ Emax™ Immunoassay Systems


Optimized for the sensitive and specific detection of a variety of neurotrophic and growth factors

Manufacturer: promega™  G7611

 View more versions of this product

Catalog No. PR-G7611

  • / Each
 

Description & Specifications

Specifications

Product Type BDNF Emax™ immunoassay system
Quantity 5 x 96 Well
Conjugate HRP
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution
Storage Requirements -20°C
Detection Method Chromogenic
Sensitivity 15.6pg/mL