Please login to your online account to display your discounted pricing

Promega™ Emax™ Immunoassay Systems


Optimized for the sensitive and specific detection of a variety of neurotrophic and growth factors

Manufacturer: promega™  G7591

 View more versions of this product

Catalog No. PR-G7591

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 96 Well
Storage Requirements -20°C
Product Type TGFb2 Emax™ immunoassay system
Conjugate HRP
Sensitivity 31pg/mL
Detection Method Chromogenic
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution