Please login to your online account to display your discounted pricing

Promega™ Emax™ Immunoassay Systems


Optimized for the sensitive and specific detection of a variety of neurotrophic and growth factors

Manufacturer: promega™  G7590

 View more versions of this product

Catalog No. PR-G7590

  • / Each
 

Description & Specifications

Specifications

Quantity 2 x 96 Well
Includes Antibodies, HRP Conjugate, standard, Buffers, TMB solution
Product Type TGFb1 Emax™ immunoassay system
Sensitivity 31pg/mL
Conjugate HRP
Detection Method Chromogenic
Storage Requirements -20°C