Please login to your online account to display your discounted pricing

Promega™ Dual-Glo™ Luciferase Assay System


Generates both firefly and Renilla luciferase luminescence signals

Manufacturer: promega™  E2980

 View more versions of this product

Catalog No. PR-E2980

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements Store Dual-Glo Substrates at -20°C; store Dual-Glo Buffers below 25°C.
Format 96 and 384-well Plates
Quantity 10 x 100mL