Please login to your online account to display your discounted pricing

Promega™ Dual-Glo™ Luciferase Assay System


Generates both firefly and Renilla luciferase luminescence signals

Manufacturer: promega™  E2940

 View more versions of this product

Catalog No. PR-E2940

  • / Each
 

Description & Specifications

Specifications

Quantity 100mL
Format 96 and 384-well Plates
Storage Requirements Store Dual-Glo Substrates at -20°C; store Dual-Glo Buffers below 25°C.