missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega DPPIV-Glo™ Protease Assay
The DPPIV-Glo™ Protease Assay is a homogeneous, luminescent assay that measures dipeptidyl peptidase IV (DPPIV) activity.
$688.35 - $1793.89
Specifications
Product Type | DDPIV-GLO Protease assay |
---|---|
Storage Requirements | -30°C to -10°C |
Includes: Part No. G8350: DPPIV-Glo™ Buffer (10mL); DPPIV-Glo Substrate (26.5μL); Luciferin Detection Reagent (1 Each)
Part No. G8351: DPPIV-Glo™ Buffer (50mL); DPPIV-Glo Substrate (115.2μL); Luciferin Detection Reagent (1 Each)
Catalog Number | Mfr. No. | Quantity | For Use With (Application) | Kit Contents | Sufficient For | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | For Use With (Application) | Kit Contents | Sufficient For | Price | Quantity | ||
PR-G8350
|
Promega
G8350 |
10 mL | Purified Enzyme preparation, DPPIV Enzyme activity determination, Kinetic studies of DPPIV inhibito r | DPPIV-Glo buffer (10 mL), DPPIV-Glo substrate (26.5 μL), Luciferin detection reagent (1 Each) | 100 Assays at 100μL/assay or 200 assays at 50μL/assay in 96-well plates or 400 assays at 25μL/assay in 384-well plate |
Each for $688.35
|
|
||
PR-G8351
|
Promega
G8351 |
50 mL | DPPIV Enzyme activity determination, Kinetic studies of DPPIV inhibito r | DPPIV-Glo buffer (50 mL), DPPIV-Glo substrate (115.2 μL), Luciferin detection reagent (1 Each) | 500 Assays at 100μL/assay or 1000 assays at 50μL/assay in 96-well plates or 2000 assays at 25μL/assay in 384-well plate |
Each for $1,793.89
|
|
||
Description
- Homogeneous, Luminescent Assay that Measures Dipeptidyl Peptidase IV Activity
- Simple add-mix-measure protocol
- Results in as little as 30 minutes after reagent addition
- Signal stability allows batch processing
Order Info
Part No. G8350 provides sufficient reagents for 100 assays at 100μL/assay or 200 assays at 50μL/assay in 96-well plates or 400 assays at 25μL/assay in 384-well plates.
Part No. G8351 provides sufficient reagents for 500 assays at 100μL/assay or 1000 assays at 50μL/assay in 96-well plates or 2000 assays at 25μL/assay in 384-well plates.
Specifications
DDPIV-GLO Protease assay | |
-30°C to -10°C |