missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ DNA-Dependent Protein Kinase DFS Item

Phosphorylates several DNA-binding substrates in vitro, including the tumor suppressor protein p53, the SV40 large T-antigen and several transcription factors

Manufacturer:  Promega™ V5811

 View more versions of this product

Catalog No. PR-V5811



Description

Description

  • Isolated from HeLa nuclear extracts as a complex consisting of a 400kDa catalytic subunit and a 155kDa heterodimeric DNA-binding component named Ku, which itself consists of subunits of approximately 85kDa and 70kDa
  • Unit Definition: One unit is defined as the amount of enzyme required to transfer 1pmol of phosphate to DNA-Dependent Protein Kinase Peptide Substrate in 1 minute at 30°C
  • Storage Buffer: 25mM HEPES (pH 7.5), 50mM KCl, 0.2mM EDTA, 10mM MgCl2, 1mM DTT, 10% glycerol

Specifications

Specifications

DNA-dependent Protein Kinase
Colorless
HeLa nuclear extracts
85 and 70
Phosphorylates several DNA-binding Substrates in-vitro, including the tumor suppressor Protein p53, the SV40 large T-antigen and several transcription factors
<−65°C
Liquid
400kDa Catalytic subunit and a 155kDa heterodimeric DNA-binding component named Ku
7.5
2,500U
25mM HEPES (pH 7.5), 50mM KCl, 0.2mM EDTA, 10mM MgCl2, 1mM DTT, 10% Glycerol
Documents
Promotions

Promotions