Please login to your online account to display your discounted pricing

Promega™ Conjugated Antibodies, Anti-ACTIVE™ Qualified


Secondary antibodies specifically tested for use with Promega Anti-ACTIVE antibodies

Manufacturer: promega™  V7951

 View more versions of this product

Catalog No. PR-V7951

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Secondary
Monoclonal or Polyclonal Polyclonal
Conjugate HRP
Host Species Donkey