Please login to your online account to display your discounted pricing

Promega™ Conjugated Antibodies, Anti-ACTIVE™ Qualified

Secondary antibodies specifically tested for use with Promega Anti-ACTIVE antibodies

$175.48 - $175.48

Specifications

Host Species Donkey
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

PR-V7951

 
promega™
V7951
Each for $175.48
Description & Specifications

Specifications

Host Species Donkey
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary