Please login to your online account to display your discounted pricing

Promega™ Conjugated Antibodies


Ideal for use in protein detection in Western Blots and ELISAs.

Manufacturer: promega™  G1351

 View more versions of this product

Catalog No. PR-G1351

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store the unopened product at –20°C. Store opened Anti-Human IgG (H+L), HRP Conjugate, Anti-Mouse IgG (H+L), HRP Conjugate and Anti-Rabbit IgG (H+L), HRP Conjugate at 4°C.
Primary or Secondary Secondary
Monoclonal or Polyclonal Polyclonal
Format Liquid
Applications Western blotting, ELISA, Dot blotting
Quantity 300μL
Regulatory Status RUO
Host Species Rabbit
Conjugate HRP

  • Ready-to-Use Formulation: No need to dissolve the antibody
  • Applications:
  • Western blotting
  • Enzyme-linked immunosorbent assay (ELISA)
  • Dot blotting