Please login to your online account to display your discounted pricing

Promega™ Conjugated Antibodies


Ideal for use in protein detection in Western Blots and ELISAs.

Manufacturer: promega™  V1151

 View more versions of this product

Catalog No. PR-V1151

  • / Each
 

Description & Specifications

Specifications

Description Donkey Anti-Goat IgG
Storage Requirements Store Anti-Mouse, Anti-Human, Anti-Rabbit (H+L) (Fc), AP conjugate antibodies at 2° to 10°C. Store Donkey Anti-goat IgG, AP at -30° to -10°C.
Regulatory Status RUO
Applications Western blotting, ELISA, Dot blotting
Quantity 60μL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary
Format Liquid
Conjugate AP
Host Species Donkey

  • Ready-to-Use Formulation: No need to dissolve the antibody
  • Applications:
  • Western blotting
  • Enzyme-linked immunosorbent assay (ELISA)
  • Dot blotting