Please login to your online account to display your discounted pricing

Promega™ Conjugated Antibodies DFS Item

Ideal for use in protein detection in Western Blots and ELISAs.

$129.47 - $200.08

Specifications

Applications Dot Blot
Applications Western Blot
Format Liquid
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
View More Specs
Products
Catalog Number Mfr. No. Description Conjugate Host Species Quantity Price Quantity    

PR-S3721

DFS Item
 
promega™
S3721
Anti-Mouse IgG (H+L) Alkaline Phosphatase Goat 100μL Each for $132.68

PR-S3831

DFS Item
 
promega™
S3831
Anti-Rat IgG (H+L) Alkaline Phosphatase Goat 100μL Each for $129.47

PR-S3731

DFS Item
 
promega™
S3731
Anti-Rabbit IgG (Fc) Alkaline Phosphatase Goat 100μL Each for $132.68

PR-S3821

DFS Item
 
promega™
S3821
Anti-Human IgG (H+L) Alkaline Phosphatase Goat 100μL Each for $132.68

PR-V1151

DFS Item
 
promega™
V1151
Donkey Anti-Goat IgG Alkaline Phosphatase Donkey 60μL Each for $175.49

PR-V8051

DFS Item
 
promega™
V8051
Donkey Anti-Goat IgG HRP Donkey 60μL Each for $180.82

PR-W4021

DFS Item
 
promega™
W4021
Anti-Mouse IgG (H+L) HRP Goat 300μL Each for $139.10

PR-W4011

DFS Item
 
promega™
W4011
Anti-Rabbit IgG (H+L) HRP Goat 300μL Each for $139.10

PR-W4031

DFS Item
 
promega™
W4031
Anti-Human IgG (H+L) HRP Goat 300μL Each for $139.10

PR-G1351

DFS Item
 
promega™
G1351
Each for $200.08
Description & Specifications

Specifications

Applications Dot Blot
Applications Western Blot
Format Liquid
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Secondary

  • Ready-to-Use Formulation: No need to dissolve the antibody
  • Applications:
  • Western blotting
  • Enzyme-linked immunosorbent assay (ELISA)
  • Dot blotting