Promega Coelenterazines
We offer Coelenterazine (native) and Coelenterazine-h. Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin.
Supplier: Promega S2001
Description
- Supplied as a dried substrate for easy preparation in methanol or ethanol
Specifications
423.5 | |
For studying cellular phenomena in expression assays in eukaryotic cells | |
Film |
-30°C to -10°C | |
1 x 250 μg |