missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Coelenterazines DFS Item

We offer Coelenterazine (native) and Coelenterazine-h. Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin.

$147.19 - $188.32

Specifications

Format Film
For Use With (Application) For studying cellular phenomena in expression assays in eukaryotic cells
Quantity 1 x 250μg
Storage Requirements −30 to −10°C
View More Specs

Products 2
Catalog Number Mfr. No. Molecular Weight Price Quantity  
Catalog Number Mfr. No. Molecular Weight Price Quantity  
PR-S2001 View Documents Promega™
S2001
423.5 Each for $188.32
 
PR-S2011 View Documents Promega™
S2011
407.5 Each for $147.19
 
Description

Description

  • Supplied as a dried substrate for easy preparation in methanol or ethanol
Specifications

Specifications

Film
For studying cellular phenomena in expression assays in eukaryotic cells
1 x 250μg
−30 to −10°C