Please login to your online account to display your discounted pricing

Promega™ CellTiter-Glo™ Luminescent Cell Viability Assay Kit DFS Item


Luminescent method for determining the number of viable cells in culture based on the amount of ATP present

Manufacturer: promega™  G7571

 View more versions of this product

Catalog No. PR-G7571

  • / Each
 

Description & Specifications

Specifications

Quantity 10 × 10mL
Sufficient For 1000 × 100μL assays or 4000 × 25μL assays
For Use With (Equipment) For 96 or 384-well plates
For Use With (Application) For automated high throughput screening, cell proliferation assays and cytotoxicity assays, determining the number of viable cells in culture based on the amount of ATP present
Storage Requirements -20°C