Please login to your online account to display your discounted pricing

Promega™ CellTiter-Fluor™ Cell Viability Assay DFS Item


Nonlytic, single-reagent-addition fluorescence assay that measures the relative number of viable cells in a population based on measurement of a conserved and constitutive protease activity within live cells

Manufacturer: promega™  G6082

 View more versions of this product

 Disclaimers

Patent Pending.

Catalog No. PRG6082

  • / Each
 

Description & Specifications

Specifications

Detection Method Fluorescence
For Use With (Equipment) 96- or 384-well plates
Storage Requirements -20°C
For Use With (Application) For normalizing data for live cell number
Quantity 2 x 50mL