Please login to your online account to display your discounted pricing

Promega™ CellTiter 96™ Nonradioactive Cell Proliferation Assay (MTT) DFS Item


Collection of reagents provides a convenient method of determining viable cell number in proliferation

Manufacturer: promega™  G4000

 View more versions of this product

Catalog No. PR-G4000

  • / Each
 

Description & Specifications

Specifications

Quantity 1000 Assays
For Use With (Equipment) For 96-well plates
For Use With (Application) For cell proliferation, cell viability, cytotoxicity, chemosensitivity, cell attachment, chemotaxis
Storage Requirements Dye solution at -20°C and solubilization/stop solution at room temperature