Please login to your online account to display your discounted pricing

Promega™ CellTiter 96™ AQueous One Solution Cell Proliferation Assay (MTS) DFS Item


Determines the number of viable cells in proliferation, cytotoxicity, or chemosensitivity assays. Absorbance of the formazan product at 490nm can be measured from the assay plate and is directly proportional to the number of living cells in culture.

Manufacturer: promega™  G3581

 View more versions of this product

Catalog No. PR-G3581

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C, protected from light.
Quantity 5000 Assays
For Use With (Equipment) For multiwell plates
For Use With (Application) For cell proliferation, cell viability, cytotoxicity, chemosensitivity, cell attachment determination, chemotaxis

  • Convenient solution combines PES, a chemically stable electron coupling reagent, and MTS, a novel tetrazolium compound that is soluble in tissue culture medium.
  • Solution is supplied filter sterilized and ready to use
  • Simple “add-incubate-measure” format enables design of homogeneous high-throughput screening assays
  • Entire assay is performed in a multiwell plate with no washing or cell harvesting; no solubilization steps required
  • Eliminates need for scintillation cocktail or volatile organic solvents; produces no radioactive waste
  • Plates can be read and returned to incubator for further color development

 

Cell proliferation; Cell viability; Cytotoxicity; Chemosensitivity; Cell attachment determination; Chemotaxis