Please login to your online account to display your discounted pricing

Promega™ Reagent Powder for CellTiter 96™ AQueous Nonradioactive Cell Proliferation Assay Kit (MTS)


Manufacturer: promega™  G1111

 View more versions of this product

Catalog No. PR-G1111

  • / Each
 

Description & Specifications

Specifications

Quantity 1g
For Use With (Application) For determining the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays, cell viability, chemotaxis
Storage Requirements -20°C
Detection Method Colorimetric