Please login to your online account to display your discounted pricing

Promega™ Cell Permeable Inhibitors

Blocks the activation of MEK1, therefore inhibiting the subsequent phosphorylation and activation of MAP kinase

$217.51 - $217.51

Specifications

For Use With (Application) Blocks the activation of MEK1
Quantity 5mg
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Kit Contents Price Quantity    

PR-V1201

 
promega™
V1201
LY 294002 Each for $217.51
Description & Specifications

Specifications

For Use With (Application) Blocks the activation of MEK1
Quantity 5mg
Storage Requirements -20°C

  • Supplied as a dried material
  • Resuspend 5mg in 935μl of DMSO to produce a stock solution of 20mM