Promega™ Caspase Inhibitors DFS Item
Solubilized in DMSO for easy addition to cell culture extracts
Manufacturer:  Promega™ G5961
 View more versions of this product
Catalog No. PR-G5961
Specifications Description & Specifications
Specifications
Ac-DEVD-CHO Caspase Inhibitor
10mM
Ac-DEVD-CHO Caspase
Inhibitor of Caspase-3/7 (DEVDase) activity
Liquid
Colorless
100μL
-20°C