Please login to your online account to display your discounted pricing

Promega™ CaspACE™ Assay System, Colorimetric DFS Item


For the qualitative or quantitative determination of caspase activity, an early indicator of apoptosis

Manufacturer: promega™  G7351

 View more versions of this product

Catalog No. PR-G7351

  • / Each
 

Description & Specifications

Specifications

Format Fluid
pH 7.5
Storage Requirements -20°C
Kit Contents 1 x 5mL Caspase Assay Buffer, 1 x 125μL Caspase inhibitor, Z-VAD-FMK, 1 x 100μL Caspase-3 substrate Ac-DEVD-pNA, 1 x 100μL 100mM pNA standard, 1 x 20mL Cell Lysis Buffer
Quantity 50 Assays