missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ CaspACE™ Assay System, Colorimetric DFS Item

For the qualitative or quantitative determination of caspase activity, an early indicator of apoptosis

$568.17 - $791.80

Specifications

Format Fluid
pH 7.5
Storage Requirements -20°C
View More Specs

Includes: 100μL Caspase-3 Substrate Ac-DEVD-pNA; 50μL Caspase Inhibitor Z-VAD-FMK; 100μL pNA Standard, 100mM; 5mL Caspase Assay Buffer; 20mL Cell Lysis Buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
PR-G7220 Promega™
G7220
1 x 5mL Caspase Assay Buffer, 1 x 125μL Caspase inhibitor, Z-VAD-FMK, 2 x 100μL Caspase-3 substrate Ac-DEVD-pNA, 1 x 100μL 100mM pNA standard, 1 x 20mL Cell Lysis Buffer 100 Assays Each for $791.80
PR-G7351 Promega™
G7351
1 x 5mL Caspase Assay Buffer, 1 x 125μL Caspase inhibitor, Z-VAD-FMK, 1 x 100μL Caspase-3 substrate Ac-DEVD-pNA, 1 x 100μL 100mM pNA standard, 1 x 20mL Cell Lysis Buffer 50 Assays Each for $568.17
Specifications

Description

Colorimetric substrate produces a yellow color that is proportional to the amount of DEVDase activity present in the sample. Kit can be used to determine total caspase-3 activity or screen for inducers or inhibitors of caspase activity.

  • Can be used with purified enzyme preparations, cell extracts, or tissue lysates
  • Contains sufficient reagents to perform 50 or 100 assays

Description & Specifications

Specifications

Fluid
7.5
-20°C