missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega CAMK4 Kinase Enzyme System DFS Item

Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family.

$615.83 - $1,230.58

Specifications

Storage Requirements COMPONENTS AT DIFFERENT TEMPS
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity  
PRV2951 View Documents Promega™
V2951
Involved in neural activity-dependent signaling in neuronal nucleus 10μg Each for $615.83
 
PRV9091 View Documents Promega™
V9091
Involved in neural activity-dependent signaling, ADP-Glo™ Kinase assay measures ADP formed from Kinase reaction 1/ea Each for $1,230.58
 
Description

Description

  • Easily Screen and Profile CAMK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

COMPONENTS AT DIFFERENT TEMPS