Promega™ CAMK4 Kinase Enzyme System DFS Item
Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family.
Manufacturer:  Promega™ V9091
 View more versions of this product
Catalog No. PRV9091
Description
Description
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
1 x 10μg CAMK4 Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay
Involved in neural activity-dependent signaling, ADP-Glo™ Kinase assay measures ADP formed from Kinase reaction
1/ea
COMPONENTS AT DIFFERENT TEMPS