Please login to your online account to display your discounted pricing

Promega™ BenchTop DNA Markers DFS Item


Ready-to-load markers offer the convenience of room temperature storage

Manufacturer: promega™  G7511

 View more versions of this product

Catalog No. PR-G7511

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
Formulation Supplied in a stabilizing solution of 1X blue/orange loading dye
Color Blue/Orange
Quantity 250μL
Format Liquid
Concentration 0.2μg/μL
Storage Requirements 22° to 25°C
No. of Bands 11