Please login to your online account to display your discounted pricing

Promega™ BenchTop DNA Markers


Ready-to-load markers offer the convenience of room temperature storage

Manufacturer: promega™  G7521

 View more versions of this product

Catalog No. PR-G7521

  • / Each
 

Description & Specifications

Specifications

No. of Bands 15
Quantity 250μL
Storage Requirements 22° to 25°C
Concentration 0.2μg/μL
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
Format Liquid
Color Blue/Orange
Concentration 0.2μg/μL
Format Liquid
Color Blue/Orange
Reference Band 15
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
Formulation Supplied in a stabilizing solution of 1X blue/orange loading dye
Formulation Supplied in a stabilizing solution of 1X blue/orange loading dye
Sufficient For 50 Lanes