Please login to your online account to display your discounted pricing

Promega™ Beetle Luciferin, Potassium Salts


Can be used as reporter genes for studying transcription regulation in transient assay systems and as markers for stably transformed eukaryotic cells

Manufacturer: promega™  E1602

 View more versions of this product

Catalog No. PR-E1602

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 50mg Beetle Luciferin, Potassium salt
Molecular Weight 318.4
Quantity 50mg
Storage Requirements -70°C
Format Anhydrous